Learn More
Invitrogen™ Ly-49I Monoclonal Antibody (YLI-90), FITC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ A15413
Product Code. 15285724
  • € 414.00 / 100µg
Description
Ly-49I Monoclonal Antibody for Flow
Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ly-49I
Monoclonal
0.5 mg/mL
PBS with <0.1% sodium azide; pH 7.4
0
Klra9
CHO-K1 cells transfected with the B6 allele of the Ly-49I gene, K1ra9.
100 μg
Primary
Mouse
Antibody
IgG1 κ
Flow Cytometry
YLI-90
FITC
Klra9
5E6; Killer cell lectin-like receptor 3; killer cell lectin-like receptor subfamily A, member 9; Klra3; Klra9; Ly-49; Ly49C; Ly-49c; Ly49I; Ly49I receptor; Ly-49I receptor; LY49I1; Lymphocyte antigen 49c; Nk2.1; T-cell surface glycoprotein Ly-49C
Mouse
Purified
RUO
16640
4°C, store in dark