missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6A/E Antibody (RM0330-2M39), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11871AF532
This item is not returnable.
View return policy
Description
Ly-6A/E Monoclonal specifically detects Ly-6A/E in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Ly-6A/E | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LY6A | |
| Mouse recombinant Ly-6A/E | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| RM0330-2M39 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Ly 6A/E, Ly6A.2, Ly6A/E, Ly6E.1, Lymphocyte antigen 6 complex locus A, Lymphocyte antigen 6 complex locus E, Retinoic acid induced gene E, RIGE, SCA1, SCA-1, SCA2, Stem cell antigen 1, Stem cell antigen 2, T cell activating protein, TAP, Thymic shared antigen 1, TSA1 | |
| Rat | |
| Protein A or G purified | |
| Hematopoietic Stem Cell Markers, Stem Cell Markers | |
| 110454.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction