Learn More
Ly-6A/E Rat anti-Mouse, FITC, Clone: E13-161.7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553335
Description
The E13-161.7 antibody reacts with Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Sca1 (Ly-6A/E), a phosphatidylinositol-anchored protein of about 18kDa, is expressed on the multipotent hematopoietic stem cell (HSC) in mice w
Host Species: Rat
Clone: E13-161.7
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB/c mouse-derived “pre-T” cell hybridoma
Flow Cytometry
Specifications
| Ly-6A/E | |
| Monoclonal | |
| 0.5mg/mL | |
| Sca-1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| E13-161.7 | |
| FITC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB/c mouse-derived “pre-T” cell hybridoma | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.