missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6A Recombinant Rabbit Monoclonal Antibody (4N5K2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538029
This item is not returnable.
View return policy
Description
Positive Samples: Mouse lung, Mouse spleen, Mouse kidney Immunogen sequence: MDTSHTTKSC LLILLVALLC AERAQGLECY QCYGVPFETS CPSITCPYPD GVCVTQEAAV IVDSQTRKVK NNLCLPICPP NIESMEILGT KVNVKTSCCQ.
LY6A is included in a family of proteins that are highly homologous, low molecular weight glycophosphotidylinositol-linked proteins expressed on hematopoietic cells and lymphoid cells.
Specifications
| Ly-6A | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05533 | |
| Ly6a | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse Ly-6A/E (Sca-1) (P05533). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4N5K2 | |
| Unconjugated | |
| Ly6a | |
| Ly6; Ly6a; Ly-6A.2; Ly-6A.2/Ly-6E.1; Ly-6A/E; Ly-6E.1; lymphocyte antigen 6 complex, locus A; lymphocyte antigen 6A-2/6E-1; Sca1; SCA-1; Stem cell antigen 1; TAP; T-cell-activating protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 110454 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction