missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6B.2 Antibody (7/4), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-13077DL594
This item is not returnable.
View return policy
Description
Ly-6B.2 Monoclonal antibody specifically detects Ly-6B.2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Ly-6B.2 | |
| Monoclonal | |
| DyLight 594 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2024-07-04 00:00:00 | |
| 50mM Sodium Borate | |
| Cultured bone marrow cells | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction