missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ly-6B.2 Monoclonal Antibody (7/4), FITC
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA516540

Product Code. 15158507

  • € 497.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse Ly-6B.2 monoclonal antibody, clone 7/4 recognizes the Ly-6B.2 antigen.

Ly-6B.2 recognizes the Ly-6B.2 antigen. Ly-6B.2 is approximately 25-30 kDa, GPI-anchored, heavily glycosylated protein expressed on neutrophils, inflammatory monocytes and some activated macrophages. High levels of expression are seen in bone marrow, spleen, lung and lymph nodes. N-glycanase treatment of thioglycollate elicited peritoneal neutrophil lysates lowers the apparent molecular weight of Ly-6B.2 to approximately 15 kDa.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ly-6B.2
Monoclonal
0.1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
0
Rat
Protein G
RUO
110453
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
7/4
FITC
Ly6b
Ly6b
Cultured bone marrow cells.
100 μg
Primary
Mouse
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.