missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6C Antibody (ER-MP20), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65413MFV450
This item is not returnable.
View return policy
Description
Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry
Specifications
| Ly-6C | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| 17067 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| ER-MP20 | |
| Flow Cytometry | |
| Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C | |
| Balb/c macrophage precursor cell hybrids | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction