missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6C Antibody (PSH04-51), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32548
This item is not returnable.
View return policy
Description
Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Western Blot
Specifications
| Ly-6C | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C | |
| Synthetic peptide within mouse Ly-6C aa 51-100/ 131. (Uniprot: P0CW02) | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| PSH04-51 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 17067 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction