missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G/Ly-6C Antibody (RM0064-6D17), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11784UV
This item is not returnable.
View return policy
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ly-6G/Ly-6C | |
| Monoclonal | |
| DyLight 350 | |
| GR, Gr-1, Ly-6, Ly6G | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This antibody detects mouse Ly-6G (Gr-1), the LY-6G/LY-6C complex. | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0064-6D17 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| LY6G | |
| Cell lysates of mouse neutrophils Ig | |
| 0.1 mL | |
| Dendritic Cell Markers, Inflammation, Myeloid Cell Markers, Myeloid derived Suppressor Cell | |
| 546644 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction