missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6G/Ly-6C Monoclonal Antibody (NIMP-R14), Biotin
Rat Monoclonal Antibody
Brand: Invitrogen™ MA549078
This item is not returnable.
View return policy
Description
The monoclonal antibody NIMP-R14 is highly specific for murine Ly-6G and Ly-6C.
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications
| Ly-6G/Ly-6C | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P0CW02, P0CW03, P35461 | |
| Ly6c1, Ly6c2, Ly6g | |
| Purified BALB/c mouse neutrophils. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| NIMP-R14 | |
| Biotin | |
| Ly6c2 | |
| AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
| Rat | |
| Protein G | |
| RUO | |
| 100041546, 17067, 546644 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction