missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Brilliant Violet™ 711, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
€ 196.00 - € 386.00
Specifications
| Antigen | Ly-6G/Ly-6C |
|---|---|
| Clone | RB6-8C5 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17805207
|
Invitrogen™
407-5931-80 |
25 μg |
€ 196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17885237
|
Invitrogen™
407-5931-82 |
100 μg |
€ 386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.Specifications
| Ly-6G/Ly-6C | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
| Ly6c1, Ly6c2, Ly6g | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| RB6-8C5 | |
| Flow Cytometry | |
| Brilliant Violet 711 | |
| Rat | |
| Mouse | |
| P0CW02, P0CW03, P35461 | |
| 100041546, 17067, 546644 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel