missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G and Ly-6C Rat anti-Mouse, BUV563, Clone: RB6-8C5, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 741226
This item is not returnable.
View return policy
Description
Ly-6G and Ly-6C Monoclonal antibody specifically detects Ly-6G and Ly-6C in Mouse samples. It is validated for Flow Cytometry
Specifications
| Ly-6G and Ly-6C | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
| Not Reported | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| RB6-8C5 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction