missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ly-6G Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5141170
This item is not returnable.
View return policy
Description
Ly-6G Polyclonal Antibody for Western Blot
Ly6G (Lymphocyte antigen 6 complex locus G6D) is a component of the myeloid differentiation antigen Gr-1, together with Ly6C. The Ly6G protein has a molecular weight of approximately 21-25 kDa and is expressed by myeloid derived cells in the bone marrow. Ly6G is a good marker for detection of peripheral neutrophils, monocytes and granulocytes. Expression of Gr-1 in bone marrow correlates with granulocyte differentiation and maturation. The physiological role of Ly6G remains unclear but studies have shown its involvement in anti-tumor response, and studies with neutrophils suggest that Ly6G ligation may have an inhibitory effect on local immune responses.
Specifications
| Ly-6G | |
| Polyclonal | |
| Unconjugated | |
| Ly6g | |
| Gr1; Gr-1; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6G | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 546644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 25% glycerol and 0.05% sodium azide; pH 7.4 | |
| P35461 | |
| Ly6g | |
| Synthetic peptide within mouse Ly-6G aa 1-50/134. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto