missing translation for 'onlineSavingsMsg'
Learn More

Ly-6G Rat, PerCP-Cy5.5, Clone: 1A8, enQuireBio™

Rat Monoclonal Antibody

€ 124.00 - € 364.00

Specifications

Antigen Ly-6G
Clone 1A8
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15932218
View Documents Promotion Details
EnquireBio
QAB56-PCP55-25UG
25 μg
€ 124.00
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15922218
View Documents Promotion Details
EnquireBio
QAB56-PCP55-100UG
100 μg
€ 364.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Ly-6G
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
546644
IgG2a κ
Protein A/G Purified
1A8
Flow Cytometry
PerCP-Cyanine5.5
RUO
XP_006521677
Ly6g
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications