missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY3177833, MedChemExpress
Description
LY3177833 is a CDC7 and pMCM2 inhibitor with IC50 values of 3.3 nM and 290 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | LY3177833 |
| Physical Form | Solid |
| CAS | 1627696-51-8 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H12FN5O |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 30 mg/mL (96.99 mM) |
| SMILES | O=C1N[C@@](C)(C2=NC=NC=C2F)C3=C1C=C(C4=CNN=C4)C=C3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?