missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly49F Antibody (HBF-719), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28098
This item is not returnable.
View return policy
Description
Ly49F Monoclonal specifically detects Ly49F in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Ly49F | |
| Monoclonal | |
| 0.5mg/mL | |
| Flow Cytometry <= 1 ug/10^6 cells | |
| Klra6 | |
| CHO-K1 cells transfected with B6 allele of Ly49F gene | |
| 0.25 mg | |
| Primary | |
| B6 allele of the Ly49F receptor | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry | |
| HBF-719 | |
| Biotin | |
| Killer cell lectin like receptor 6, Klra6, Ly49F antigen, Lymphocyte antigen 49F, T cell surface glycoprotein Ly 49F | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 16637 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction