missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LY6G5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570566
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 93%; Horse: 79%; Human: 100%; Pig: 79%.
LY6G5B belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
Specifications
| LY6G5B | |
| Polyclonal | |
| Unconjugated | |
| LY6G5B | |
| C6orf19; G5B; LY6G5B; Lymphocyte antigen 6 complex locus protein G5b; lymphocyte antigen 6 complex, locus G5B; lymphocyte antigen 6 family member G5B; lymphocyte antigen-6 G5B splicing isoform 288; lymphocyte antigen-6 G5B splicing isoform 452; lymphocyte antigen-6 G5B splicing isoform 690; lymphocyte antigen-6 G5B splicing isoform 837 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 58496 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NDX9 | |
| LY6G5B | |
| synthetic peptide directed towards the middle region of human LY6G5B (aa 60-109). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction