missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LY6G6D Monoclonal Antibody (3D6E10)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA548776

Product Code. 17966111

  • € 512.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LY6G6D Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA

G6D, also known as Lymphocyte antigen 6 complex locus G6D (LY6G6D), is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, that is comprised of cell-surface proteins involved in the immune system and cellular recognition. It is a disulfide linked homodimer with an Ig-like V-type domain and is known to interact with GRB2 and GRB7 in a phosphorylation-dependent manner thus playing an important role in their downstream signal transduction pathways.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LY6G6D
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
O95868, Q6MG58, Q9Z1Q3
LOC102119090, LY6G6D
Purified recombinant fragment of human G6D (AA: 20-104) expressed in E. Coli.
100 μg
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG1
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
3D6E10
Unconjugated
LY6G6D
C6orf23; DADB-110M10.6; G6D; LY6-D; LY6G6D; Lymphocyte antigen 6 complex locus protein G6d; lymphocyte antigen 6 complex, locus G6D; lymphocyte antigen 6 family member G6D; lymphocyte antigen-6 G6D; megakaryocyte-enhanced gene transcript 1 protein; MEGT1; NG25; Protein Ly6-D
Mouse
Protein G
RUO
102119090, 114654, 415062, 58530
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.