missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6K Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24405JF549
This item is not returnable.
View return policy
Description
Ly6K Polyclonal antibody specifically detects Ly6K in Human samples. It is validated for Western Blot, ELISA
Specifications
| Ly6K | |
| Polyclonal | |
| Western Blot, ELISA | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| Reacts with residues 82-100 RQRDPEDSQRTDEGDNRVW of the 14 kDa human Ly-6K protein. | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 54742 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction