missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LY9 Monoclonal Antibody (OTI2H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811828
This item is not returnable.
View return policy
Description
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.Specifications
| LY9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HBG7 | |
| LY9 | |
| Human recombit protein fragment corresponding to amino acids 48-454 of human LY9 produced in SF9 cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2H4 | |
| Unconjugated | |
| LY9 | |
| CD229, SLAMF3, hly9, mLY9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction