Learn More
Lycopene (>80%), TRC
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Specifications
Specifications
| Chemical Name or Material | Lycopene |
| CAS | 502-65-8 |
| Molecular Formula | C40 H56 |
| Quantity | 10 mg |
| Synonym | ψ,ψ-Carotene, Lycopene, all-trans- (8CI), (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-Octamethyl-2,6,8,10,12,14,16,18,20,22,24,26,30-dotriacontatridecaene, (all-E)-2,6,10,14,19,23,27,31-Octamethyl-2,6,8,10,12,14,16,18,20,22,24,26,30-dotriacontatridecaene, (all-E)-Lycopene, 2,6,8,10,12,14,16,18,20,22,24,26,30-Dotriacontatridecaene, 2,6,10,14,19,23,27,31-octamethyl-, (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-, C.I. 75125, E 160d, Lyco Vit, Lyconat, Lycopene, Lycopene 7, Lycopene Base, Lycored, Mexoryl SAQ, NSC 407322, Redivivo, Solanorubin, Tomat-O-Red, all-trans-Lycopene, trans-Lycopene |
| InChI Formula | InChI=1S/C40H56/c1-33(2)19-13-23-37(7)27-17-31-39(9)29-15-25-35(5)21-11-12-22-36(6)26-16-30-40(10)32-18-28-38(8)24-14-20-34(3)4/h11-12,15-22,25-32H,13-14,23-24H2,1-10H3/b12-11+,25-15+,26-16+,31-17+,32-18+,35-21+,36-22+,37-27+,38-28+,39-29+,40-30+ |
| SMILES | CC(=CCC\C(=C\C=C\C(=C\C=C\C(=C\C=C\C=C(/C)\C=C\C=C(/C)\C=C\C=C(/C)\CCC=C(C)C)\C)\C)\C)C |
| IUPAC Name | (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene |
| Molecular Weight (g/mol) | 536.87 |
| Formula Weight | 536.4382 |