missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYK5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99336-100ul
This item is not returnable.
View return policy
Description
LYK5 Polyclonal antibody specifically detects LYK5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LYK5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| LYK5FLJ90524, NY-BR-96, protein kinase LYK5, Serologically defined breast cancer antigen NY-BR-96, STE20-like pseudokinase, STE20-related adapter protein, STE20-related adaptor protein, STE20-related kinase adapter protein alpha, STE20-related kinase adaptor alpha, Stlk, STRAD alpha, STRADPMSE | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human LYK5. | |
| 100 μg | |
| Signal Transduction | |
| 92335 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction