missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LYK5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528594
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HepG2, mouse brain. Predicted reactivity: Human (98%), Mouse (95%), Rat (94%), Chicken (90%), Sheep (95%), Chimpanzee (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LYK5; STE20-related Adaptor Protein (STRAD); FLJ90524. Gene Map Locus is 17q23.3 and members of STE-20 like kinase family are known to stimulate MAPK pathways by directly activating MAPKKK. STRAD is a novel pseudokinase member of this family consisting of a STE-20 like kinase domain but lacks several residues that are required for its catalytic activity. It specifically binds LKB1 and plays a key role in regulating tumor suppressor activities of LKB1. It functions as an upstream activator of LKB1 and also directs the sub-cellular localization of LKB1 by anchoring it in the cytoplasm. STRAD-LKB1 interaction results in phosphorylation of STRAD and enhanced autophosphorylation of LKB1.
Specifications
| LYK5 | |
| Polyclonal | |
| Unconjugated | |
| STRADA | |
| 2610019A05Rik; 6030402H20Rik; AI480680; E130112C09Rik; LYK5; NY-BR-96; PMSE; protein kinase LYK5; serologically defined breast cancer antigen NY-BR-96; STE20-like pseudokinase; STE20-related adapter protein; STE20-related kinase adapter protein alpha; STE20-related kinase adaptor alpha; Stlk; Strad; STRAD alpha; Strada | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 72149, 92335 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q3UUJ4, Q7RTN6 | |
| STRADA | |
| Recombinant protein encompassing a sequence within the center region of human LYK5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction