Learn More
PeproTech Lymphotactin Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P40-100UG
Description
AA Sequence of recombinant protein: GSEVSDKRTC VSLTTQRLPV SRIKTYTITE GSLRAVIFIT KRGLKVCADP QATWVRDVVR SMDRKSNTRN NMIQTKPTGT QQSTNTAVTL TG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Lymphotactin (XCL1). Anti-Human Lymphotactin (XCL1)-specific antibody was purified by affinity chromatography employing an immobilized Human Lymphotactin (XCL1) matrix. Sandwich ELISA: To detect Human Lymphotactin (XCL1) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Lymphotactin (XCL1) (500-P40Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Lymphotactin (XCL1). Western Blot: To detect hLymphotactin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hLymphotactin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
Specifications
| Lymphotactin | |
| Polyclonal | |
| Unconjugated | |
| Xcl1 | |
| AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM1alpha; SCM-1alpha; SCM-1-alpha; SCM1beta; SCM-1beta; SCM-1-beta; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6375 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P47992 | |
| Xcl1 | |
| E.coli-derived Recombinant Human Lymphotactin (XCL1). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.