missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (MM0461-3T24), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11947H
This item is not returnable.
View return policy
Description
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot.
Specifications
| Lymphotoxin beta R/TNFRSF3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| LTBR | |
| Human recombinant Lymphotoxin beta R/TNFRSF3 (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0461-3T24 | |
| Western Blot | |
| CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Cytokine Research | |
| 4055 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction