missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lymphotoxin beta R/TNFRSF3 Antibody (OTI3G10), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72551B
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Lymphotoxin beta R/TNFRSF3 Monoclonal specifically detects Lymphotoxin beta R/TNFRSF3 in Human samples. It is validated for Western Blot.
Specifikationer
| Lymphotoxin beta R/TNFRSF3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LTBR | |
| Recombinant protein fragment corresponding to amino acids 31-227 of human Lymphotoxin beta R/TNFRSF3 (NP_002333) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI3G10 | |
| Western Blot | |
| CD18, D12S370, ltbetar, LT-BETA-R, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cytokine Research | |
| 4055.0 | |
| Store at 4°C in the dark. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering