Learn More
Invitrogen™ Lymphotoxin beta Receptor Monoclonal Antibody (eBio3C8 (3C8)), Functional Grade, eBioscience™

Rat Monoclonal Antibody
Brand: Invitrogen™ 16-5671-82
Description
Description: The antibody reacts with mouse lymphotoxin-beta receptor (LTBR). Northern blot analysis of tissues from adult mice showed that expression levels of LTBR mRNA were strong in lung, liver, and kidney, moderate in heart and testes, but weak in brain, thymus, spleen, and lymph nodes. The tumor necrosis factor receptor-related protein is the human receptor for the heterotrimer of lymphotoxinalpha and lymphotoxin beta. This LT alpha/LTbeta heterotrimer (LT alpha 1/beta 2) is assumed to take part in immunologic reactions by cell-cell contact, but does not bind to either TNFR1 (CD120 alpha) or TNFR2 (CD120 beta). LTBR is expressed by day 7 embryos, so it has been speculated that the LT alpha/LT beta receptor system may also have some function in early embryogenesis. Applications Reported: This eBio3C8 (3C8) antibody has been reported for use in flow cytometric analysis and to have agonistic activity (via activation of NK kappa B). Applications Tested: This eBio3C8 (3C8) antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration:...
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
Specifications
| Lymphotoxin beta Receptor | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P50284 | |
| Ltbr | |
| Affinity chromatography | |
| RUO | |
| 17000 | |
| 4°C | |
| Liquid |
| Flow Cytometry, Functional Assay | |
| eBio3C8 (3C8) | |
| Functional Grade | |
| Ltbr | |
| AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.