missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPD6 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98014DL594
This item is not returnable.
View return policy
Description
LYPD6 Polyclonal specifically detects LYPD6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| LYPD6 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| LY6/PLAUR domain containing 6, UNQ3023/PRO9821 | |
| Produced in rabbits immunized with E. coli-derived Human LYPD6 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 130574 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction