missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPD8 Antibody (961703) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9087JF635
This item is not returnable.
View return policy
Description
LYPD8 Monoclonal antibody specifically detects LYPD8 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| LYPD8 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 961703 | |
| 50mM Sodium Borate | |
| Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82 | |
| RUO | |
| 646627 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction