missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPD8 Antibody (961703) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9087P
This item is not returnable.
View return policy
Description
LYPD8 Monoclonal antibody specifically detects LYPD8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| LYPD8 | |
| Monoclonal | |
| PE | |
| PBS | |
| Human embryonic kidney cell line HEK293-derived transfected with human LYPD8, Met1-Asn215, Accession # Q6UX82 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 961703 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 646627 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction