missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYRIC Antibody (OTI1B6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72552AF405
This item is not returnable.
View return policy
Description
LYRIC Monoclonal specifically detects LYRIC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| LYRIC | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MTDH | |
| Recombinant protein fragment corresponding to amino acids 70-582 of human LYRIC (NP_848927)produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B6 | |
| Western Blot, Immunohistochemistry | |
| 3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 92140.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction