missing translation for 'onlineSavingsMsg'
Learn More

LYRIC Antibody (OTI1B6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72552

Product Code. 18111162

  • ONLINE EXCLUSIVE PRICE
    € 684.00
    € 646.80 / 100µg
    Save € 37.20
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LYRIC Monoclonal specifically detects LYRIC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LYRIC
Monoclonal
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
3D3, AEG-13D3/LYRIC, AEG1LYRIC/3D3, Astrocyte elevated gene-1 protein, LYRICMetastasis adhesion protein, Lysine-rich CEACAM1 co-isolated protein, metadherin, protein LYRIC
Mouse
63.7 kDa
100 μg
Primary
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunohistochemistry
OTI1B6
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
MTDH
Human recombinant protein fragment corresponding to amino acids 70-582 of human MTDH (NP_848927)produced in SF9 cell.
Affinity Purified
Phospho Specific
92140
Human, Mouse, Rat
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.