Learn More
DiscoverX [Lys8,9]-Neurotensin (8-13)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
[Lys8,9]-Neurotensin (8-13) is the smallest active fragment of Neurotensin, a 13 amino acid neuropeptide of the CNS that is implicated in luteinizing hormone (LH) and dopamine regulation, and prolactin release. Effects of Neurotensin activity include vasodilation, analgesia, hypothermia, and increased locomotion. This [Lys8,9]-analog of neurotensin enhances amiloride-sensitive Na+-dependent transmembrane H+ flux. Provided as 5mg of dry powder.
Specifications
Specifications
| Product Type | Control Ligands |
| Quantity | 5 mg |