Learn More
DiscoverX [Lys8,9]-Neurotensin (8-13)
Product Code. 17322399
Encompass
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 17322399
Brand: DiscoverX 921059
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
[Lys8,9]-Neurotensin (8-13) is an agonist for neurotensin receptors. Provided as 5mg of dry powder.
[Lys8,9]-Neurotensin (8-13) is the smallest active fragment of Neurotensin, a 13 amino acid neuropeptide of the CNS that is implicated in luteinizing hormone (LH) and dopamine regulation, and prolactin release. Effects of Neurotensin activity include vasodilation, analgesia, hypothermia, and increased locomotion. This [Lys8,9]-analog of neurotensin enhances amiloride-sensitive Na+-dependent transmembrane H+ flux. Provided as 5mg of dry powder.
Specifications
Product Type Control Ligands
Quantity 5 mg