Learn More
R&D Systems™ Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 10765 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-97310. For further blocking peptide related protocol, click here . |
| Gene Alias | CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 |
| Gene Symbol | KDM5B |
| Quantity | 0.1 mg |
| Research Category | Cancer, Epigenetics |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |