Learn More
MP Biomedicals™ Lysing Matrix A, Bulk
Product Code. 15762282 Shop All MP Biomedicals Products
Click to view available options
Quantity:
200 g
500 g
Unit Size:
200g
500g
Product Code. 15762282
Brand: MP Biomedicals™ 116540423
Lysing Matrix A, Bulk contains sharp, angular flakes for grinding a range of sample types for DNA isolation.
Lysing Matrix A, bulk contains a garnet matrix made of sharp, angular flakes that are chemically inert and will not bind nucleic acid. The garnet matrix is a component of Lysing Matrix A tubes. For use on FastPrep instruments or any bead beater.
  • Used to lyse, grind or homogenize tough and resistant samples
  • Results in higher lysing efficiency for tough samples due to sharp, angular and irregular surfaces
  • Garnet matrix can be used very effectively for DNA isolation, but is very agressive and should be used with caution for RNA and protein isolation applications
  • Sample types: Animal and plant tissues, insects, nematodes and lysis of bacteria, yeast, fungi, and algae
TRUSTED_SUSTAINABILITY
Specifications
Bead Type Garnet Flakes
Sample Type Animal Tissues, Plants, Insects, Worms, Bacteria, Yeast, Fungi, Algae
For Use With (Equipment) FastPrep instruments or other bead beaters
For Use With (Application) Nucleic Acid Isolation and Purification, Homogenization
Matrix Lysing Matrix A (Garnet)
Packaging Type Bulk
Product Type Lysing Matrix A (garnet)
Quantity 200 g
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.