Learn More
MP Biomedicals™ Lysing Matrix M, 4.5mL Tube
MP Biomedicals™ Lysing Matrix M, 4.5mL Tube allows for lysing of any kind of difficult to homogenize samples for DNA, RNA and Protein Extractions using the BigPrep™ adapter
Brand:  MP Biomedicals™ 116949025
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 15732462
  • € 125.00 / Pack of 25
Estimated Shipment
12-01-2021
Add to basket
Description
Description
  • Universal grinding
  • Homogenization and Lysing Matrix
  • Lyse any kind of difficult to homogenize samples for DNA, RNA and protein extractions using the BigPrep™ adapter
Certifications
DNase and Rnase free
Specifications
Specifications
Sample preparation, Sample lysis, Nucleic acid isolation and purification, Molecular biology, Gene expression, Microbiological analysis; Gene function, Gene regulation
Tough tissues, Skeletal muscle, Lung, Heart, Bone, Seeds, Spores, Plants, Fixed Samples, Food, Roots
Lysis Matrix
Room temperature
FastPrep-24, FastPrep 24 5G, FastPrep-96
M
25 x 4.5mL