missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Lysipressin

Product Code. 16450396
Encompass_Preferred
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 16450396

Brand: MedChemExpress HYP0004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lysipressin is Antidiuretic hormone that have been found in pigs and some marsupial families. Induces contraction of the rabbit urinary bladder smooth muscle, activate adenylate-cyclase.

Specifications

CAS 50-57-7
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1056.22 g/mol
Product Type Natural Products
Quantity 2 mg
Shipping Condition Room temperature
Solubility H2O : 100 mg/mL (94.68 mM∣Need ultrasonic)
Synonym Lysine vasopressin∣ [Lys8]-Vasopressin
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.