missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYSMD4 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00145748-B01P
This item is not returnable.
View return policy
Description
LYSMD4 Polyclonal antibody specifically detects LYSMD4 in Human samples. It is validated for Western Blot
Specifications
| LYSMD4 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| FLJ33008, lysM and putative peptidoglycan-binding domain-containing protein 4, LysM, putative peptidoglycan-binding, domain containing 4, MGC99501 | |
| LYSMD4 (NP_689662.2, 1 a.a. - 297 a.a.) full-length human protein. MAVGTRGTLLKKSLTVWFCGPGARSATRAVSTSLPRREQVTWCCCSGSWPRRTASTSWRCSMAANFCRHTVWLTEHGLKNEACPTKTGIDGVGFLVADIKKVNNFIREQDLYALKSVKIPVRNHGILMETHKELKPLLSPSSETTVTVELPEADRAGAGTGAQAGQLMGFFKGIDQDIERAVQSEIFLHESYCMDTSHQPLLPAPPKTPMDGADCGIQWWNAVFIMLLIGIVLPVFYLVYFKIQASGETPNSLNTTVIPNGSMAMGTVPGQAPRLAVAVPAVTSADSQFSQTTQAGS | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NP_689662.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 145748 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction