missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYSMD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62190
This item is not returnable.
View return policy
Description
LYSMD4 Polyclonal antibody specifically detects LYSMD4 in Human samples. It is validated for Western Blot
Specifications
| LYSMD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| FLJ33008, lysM and putative peptidoglycan-binding domain-containing protein 4, LysM, putative peptidoglycan-binding, domain containing 4, MGC99501 | |
| Synthetic peptides corresponding to LYSMD4(LysM, putative peptidoglycan-binding, domain containing 4) The peptide sequence was selected form the N terminal of LYSMD4. Peptide sequence PRREQVTWCCCSGSWPRRTASTSWRCSMAANTFYFRPNGAGDTRQNLIPD. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| A6NII6 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 145748 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction