missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysozyme Antibody (OTI1C9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72557AF488
This item is not returnable.
View return policy
Description
Lysozyme Monoclonal specifically detects Lysozyme in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Lysozyme | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LYZ | |
| Full length recombinant protein of human Lysozyme (NP_000230) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1C9 | |
| Western Blot, Immunohistochemistry | |
| EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis | |
| Mouse | |
| Affinity Purified | |
| Cardiovascular Biology, Immunology | |
| 4069.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction