missing translation for 'onlineSavingsMsg'
Learn More

Lysozyme Antibody (OTI1C9), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-72557CL1

Product Code. 30153258

  • € 1150.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Lysozyme Monoclonal antibody specifically detects Lysozyme in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lysozyme
Monoclonal
CoraFluor 1
EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Full length human recombinant protein of human LYZ(NP_000230) produced in HEK293T cell.
0.1 mL
Cardiovascular Biology, Immunology
4069
Store at 4°C in the dark. Do not freeze.
IgG2b
Immunohistochemistry, Western Blot
OTI1C9
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.