missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lysozyme Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128894
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17710674 200 μL 200µL
17763184 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17710674 Supplier Invitrogen™ Supplier No. PA5119264

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lysozyme Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta[1-4]glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the anti-microbial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. Missense mutations in LYZ have been identified in heritable renal amyloidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysozyme
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Lyz1
Gene Accession No. P04421, P80189, Q06283
Gene Alias 1,4-beta-N-acetylmuramidase C; 1700038F02Rik; 4-beta-N-acetylmuramidase C; AI326280; Allergen Gal d IV; bA534G20.1; c-type lysozyme; dystrophin; egg white lysozyme; Gal d 4; KAAG648; LYC2; Lys; Lysm; lysozyme; lysozyme (renal amyloidosis); lysozyme 1; lysozyme 2; lysozyme C; Lysozyme C type M; lysozyme C type P; Lysozyme C, spleen isozyme; lysozyme C-1; Lysozyme C-2; lysozyme C-3; lysozyme d1; lysozyme F1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; Lysz; LYZ; Lyz1; Lyz2; LYZC; LYZD1; LYZF1; LYZL1; Lyzs; Lzm; Lzm-s1; Lzp; Lzp-s; P lysozyme structural; PRO1278; renal amyloidosis; RGD1559869; unnamed protein product; UNQ648/PRO1278
Gene Symbols LYZ, Lyz1, Lyz2
Host Species Rabbit
Immunogen Recombinant protein Lysozyme. The antigen corresponds to amino acid range 18-147 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280849, 281287, 777776, 781349
Target Species Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.