missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody (OTI8B2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72558AF532
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LOXL2 | |
| Recombinant protein fragment corresponding to amino acids 152-450 of human Lysyl Oxidase Homolog 2/LOXL2 (NP_002309) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry | |
| OTI8B2 | |
| Immunohistochemistry | |
| EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| Mouse | |
| 0.1 mL | |
| 4017 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction