Learn More
Lysyl Oxidase Homolog 2/LOXL2 Antibody (simtuzumab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28051JF669
Product Code. 30178130
  • € 419.00 / 0.10mL
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lysyl Oxidase Homolog 2/LOXL2
Monoclonal
Janelia Fluor 669
EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14
LOXL2
0.1 mL
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
4017
Store at 4°C in the dark.
IgG4
Flow Cytometry, ELISA, Functional Assay
simtuzumab
50mM Sodium Borate
Human
Protein A purified