missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE-1 Antibody (223322), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2125CL1
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal antibody specifically detects LYVE-1 in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Blockade of Receptor-ligand Interaction,Immunocytochemistry,CyTOF
Specifications
| LYVE-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1, UNQ230/PRO263, XLKD1 | |
| BaF/3 mouse pro-B cell line transfected with mouse LYVE-1, Ala24-Thr234, Accession # Q8BHC0 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
| 10894 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Western Blot, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
| 223322 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction