missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYZL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41184
This item is not returnable.
View return policy
Description
LYZL1 Polyclonal specifically detects LYZL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| LYZL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LYZL1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human. The immunogen is located within the first 50 amino acids of LYZL1. Amino Acid Squence: SADSTEKSASGAGTRNL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least two alternatively spliced transcript variants have been observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA | |
| EC 3.2.1.17, KAAG648, LYC2bA534G20.1, lysozyme-like 1, lysozyme-like protein 1, MGC33408, PRO1278 | |
| Rabbit | |
| 21 kDa | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 84569 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction