missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYZL1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41184UV
This item is not returnable.
View return policy
Description
LYZL1 Polyclonal antibody specifically detects LYZL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| LYZL1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 350 | |
| EC 3.2.1.17, KAAG648, LYC2bA534G20.1, lysozyme-like 1, lysozyme-like protein 1, MGC33408, PRO1278 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human. The immunogen is located within the first 50 amino acids of LYZL1. Amino Acid Squence: SADSTEKSASGAGTRNL | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84569 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction