missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Antibody (OTI2D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45704
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
LZIC Monoclonal specifically detects LZIC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| LZIC | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| leucine zipper and CTNNBIP1 domain containing, Leucine zipper and CTNNBIP1 domain-containing protein, Leucine zipper and ICAT homologous domain-containing protein, leucine zipper domain and ICAT homologous domain containing, MGC15436, protein LZIC | |
| Mouse | |
| 21.3 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 84328 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI2D4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| LZIC | |
| Full length human recombinant protein of human LZIC(NP_115744) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction