missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Antibody (OTI3A7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72559JF646
This item is not returnable.
View return policy
Description
LZIC Monoclonal specifically detects LZIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| LZIC | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LZIC | |
| Recombinant protein fragment corresponding to amino acids 1-190 of human LZIC (NP_115744) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3A7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| leucine zipper and CTNNBIP1 domain containing, Leucine zipper and CTNNBIP1 domain-containing protein, Leucine zipper and ICAT homologous domain-containing protein, leucine zipper domain and ICAT homologous domain containing, MGC15436, protein LZIC | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 84328.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction