missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Antibody (OTI3A7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72559PE
This item is not returnable.
View return policy
Description
LZIC Monoclonal antibody specifically detects LZIC in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| LZIC | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI3A7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| leucine zipper and CTNNBIP1 domain containing, Leucine zipper and CTNNBIP1 domain-containing protein, Leucine zipper and ICAT homologous domain-containing protein, leucine zipper domain and ICAT homologous domain containing, MGC15436, protein LZIC | |
| Human recombinant protein fragment corresponding to amino acids 1-190 of human LZIC (NP_115744) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 84328 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction